ContactUsLinkComponent
BioClean™ Vijon™ Sterile Single Use Goggles, Ansell
  75839-022
undefined
BioClean™ Vijon™ Sterile Single Use Goggles, Ansell
  75839-022
 :  BVGS
  • Description:
    Vijon™ sterile single use goggles
  • Cat. No.:
    75839-022
  • Supplier no.:
    BVGS
BioClean™ Vijon™ sterile single use goggles are lightweight and non-linting.
  • Lightweight PVC frame
  • Indirect ventilation system to maintain user comfort
  • Optically correct lens
  • Toughened polycarbonate, anti-fog, and anti-scratch lens
  • Non-linting, latex-free head band offering unrivaled comfort
  • ISO Class 4 compatible
BioClean™ Vijon™ sterile single use goggles are constructed from lightweight, ultra-soft PVC. They may also be worn over eyeglasses.
 : Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Conforms to Category 2 - Personal Protective Equipment PPE Directive 89/686/EEC.
Complies with requirements of EN 166:2001 standard for personal eye protection.
Tested to ANSI/ISEA Z87.1-2010, the American National Standard for Occupational and Educational Personal Eye and Face Protection Devices.
 : Goggles are packaged one piece per sealed PE inner bag; 10 inner bags per sealed outer PE bag; six outer bags per lined carton (60 pieces).