Q-BOND and U-BOND Column Test Mix, Restek
Catalog # 75835-356
Supplier: Restek
CAS Number:
Specifications
- Concentration:0.1% vol/vol each in heptane
- Description:Q-BOND and U-BOND Column Test Mix
- Size:1 ml/Ampule
- Cat. No.:75835-356
- Supplier no.:35202
Specifications
About this item
7 components