Order Entry
Puerto Rico
ContactUsLinkComponent
NPI-1000 Column Test Mix (8 Components), Restek
NPI-1000 Column Test Mix (8 Components), Restek
Catalog # 75835-360
Supplier:  Restek
CAS Number:  
NPI-1000 Column Test Mix (8 Components), Restek
Catalog # 75835-360
Supplier:  Restek
Supplier Number:  35224
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml each in methylene chloride
  • Description:
    NPI-1000 Column Test Mix
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-360
  • Supplier no.:
    35224

Specifications

About this item

The mix consists of (C9:0) methyl nonanoate, (C13) n-tridecane, (C15) n-pentadecane, acenaphthylene, 4-chlorophenol, 1,6-hexanediol, 4-propylaniline and 1-undecanol.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.