Order Entry
Puerto Rico
ContactUsLinkComponent
RapidVap® Trapping Valve, Labconco®
RapidVap® Trapping Valve, Labconco®
Catalog # 75841-004
Supplier:  Labconco
undefined
RapidVap® Trapping Valve, Labconco®
Catalog # 75841-004
Supplier:  Labconco
Supplier Number:  7348000

Specifications

  • Description:
    Trapping Valve
  • Size:
    12.2 W x 14.7 D x 13.7 H cm (4.8 x 5.8 x 5.4")
  • Cat. No.:
    75841-004
  • For use with:
    RapidVap® Vacuum Systems
  • Supplier no.:
    7348000

Specifications

About this item

The RapidVap® Trapping Valve aids in solvent recovery for RapidVap® Vacuum Evaporation Systems.

  • Cold Trap is required (not included)
  • Includes tubing and clamps for connection to Vacuum System
  • Compatible with RapidVap® Vacuum Systems manufactured after September 2016
Ordering information: RapidVap® Vacuum System and Cold Trap each sold separately.