Order Entry
Puerto Rico
ContactUsLinkComponent
GC Multiresidue Pesticide Standard #7, (10 components), Restek
GC Multiresidue Pesticide Standard #7, (10 components), Restek
Catalog # 75835-058
Supplier:  Restek
CAS Number:  
GC Multiresidue Pesticide Standard #7, (10 components), Restek
Catalog # 75835-058
Supplier:  Restek
Supplier Number:  32569
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml each in toluene
  • Description:
    GC Multiresidue Pesticide Standard #7
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-058
  • Supplier no.:
    32569

Specifications

About this item

The multiresidue accurately identify and quantify pesticide residues by gc-ms/ms in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.

  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Formulated and grouped for maximum long-term stability

The multiresidue pesticide standard consist of ten components acequinocyl, bromopropylate, carfentrazone ethyl, chlorobenzilate, chlorpropham, chlozolinate, DCPA methyl ester (chlorthal-dimethyl), fluazifop-p-butyl, metalaxyl and 2-phenylphenol.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Ordering information: * NOTE: When combining a large number of compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.