Order Entry
Puerto Rico
ContactUsLinkComponent
 
GC Multiresidue Pesticide Standard #4 (28 components), Restek
  75835-052
 :  Restek
 :  
GC Multiresidue Pesticide Standard #4 (28 components), Restek
  75835-052
 :  Restek
 :  32566
 :  

 

  • Concentration:
    100 µg/ml each in toluene
  • Description:
    GC Multiresidue Pesticide Standard #4
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-052
  • Supplier no.:
    32566

 

 

Organonitrogen compounds that has accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.

Contains: Acetochlor, alachlor, allidochlor, clomazone (command), cycloate, diallate(cis & trans), dimethachlor, diphenamid, fenpropathrin, fluquinconazole, flutolanil, linuron, metazachlor, methoxychlor, metolachlor, N-(2,4-dimethylphenyl)formamide, norflurazon, oxadiazon, pebulate, pretilachlor, prochloraz, propachlor, propanil, propisochlor, propyzamide, pyridaben, tebufenpyrad, triallate.

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
 : * NOTE: When combining a large number of compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.