Order Entry
Puerto Rico
ContactUsLinkComponent
LC Multiresidue Pesticide Kit, Restek
LC Multiresidue Pesticide Kit, Restek
Catalog # 75834-882
Supplier:  Restek
CAS Number:  
LC Multiresidue Pesticide Kit, Restek
Catalog # 75834-882
Supplier:  Restek
Supplier Number:  31971
CAS Number:  

Specifications

  • Description:
    LC Multiresidue Pesticide Kit
  • Size:
    Contains 1 ml each of these mixtures
  • Cat. No.:
    75834-882
  • Supplier no.:
    31971

Specifications

About this item

Accurately detect and quantify pesticides of global food safety concern in a wide range of fruits, vegetables, and other commodities by LC-MS/MS.

  • Quantitatively tested to confirm composition, detailed support documentation provided

Formulated and grouped for maximum long-term stability* and well-balanced chromatographic performance.

Optimized multiresidue pesticide method is offered free of charge and downloadable XLS file includes conditions and transition tables.

Contains 1 ml each of the following mixtures, LC multiresidue pesticide standard #1, LC multiresidue pesticide standard #2, LC multiresidue pesticide standard #3, LC multiresidue pesticide standard #4, LC multiresidue pesticide standard #5, LC multiresidue pesticide standard #6, LC multiresidue pesticide standard #7, LC multiresidue pesticide standard #8, LC multiresidue pesticide standard #9 and LC multiresidue pesticide standard #10.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: The complete kit contains 204 compounds of interest, covering many LC-determined pesticides listed by government agencies, individual ampoules also sold separately.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 204 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.