Order Entry
Puerto Rico
ContactUsLinkComponent
Methapyrilene Standard, Restek
Methapyrilene Standard, Restek
Catalog # 75835-022
Supplier:  Restek
CAS Number:  
Methapyrilene Standard, Restek
Catalog # 75835-022
Supplier:  Restek
Supplier Number:  32460
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml in methylene chloride
  • CAS No.:
    135-23-9
  • Description:
    Methapyrilene hydrochloride
  • Size:
    1 ml/Ampule
  • Formula:
    C₁₄H₂₀ClN₃S
  • Description:
    Methapyrilene hydrochloride
  • Cat. No.:
    75835-022
  • Supplier no.:
    32460

Specifications

About this item