NJDEP EPH 10/08 Rev. 2 Aromatics Fractionation Check Mix (18 Components), Restek
Catalog # 75834-486
Supplier: Restek
CAS Number:
Specifications
- Concentration:400 µg/ml each in hexane:toluene (50:50) 5 ml/Ampule
- Description:NJDEP EPH 10/08 Rev.2 Aromatics Fractionation Check Mix
- Size:5 ml/Ampule
- Cat. No.:75834-486
- Supplier no.:30545
Specifications
About this item
Consists of 18 components, those are acenaphthene, acenaphthylene, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, chrysene, dibenzanthracene, fluoranthene, fluorene, indenopyrene, 2-methylnaphthalene, naphthalene, phenanthrene, 1,2,3-trimethylbenzene and pyrene
16 components
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.