NJDEP EPH 10/08 Rev. 2 Aliphatics Matrix Spike Mix (18 components), Restek
Catalog # 75834-480
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in pentane 5 ml/Ampule
- Description:NJDEP EPH 10/08 Rev. 2 Aliphatics Matrix Spike Mix
- Size:5 ml/Ampule
- Cat. No.:75834-480
- Supplier no.:30542
Specifications
About this item
Consists of 18 components, those are n-nonane, n-decane, n-dodecane, n-tetradecane, n-hexadecane, n-octadecane, n-eicosane, n-heneicosane, n-docosane, n-tetracosane, n-hexacosane, n-octacosane, n-triacontane, n-dotriacontane, n-tetratriacontane, n-hexatriacontane, n-octatriacontane and n-tetracontane.
18 components
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.