Order Entry
Puerto Rico
ContactUsLinkComponent
Pyrene, Restek
Pyrene, Restek
  75835-654
 :  Restek
 :  
Pyrene, Restek
  75835-654
 :  Restek
 :  31282
 :  

 

  • Concentration:
    1000 µg/ml in methanol
  • CAS No.:
    129-00-0
  • Description:
    Pyrene
  • Size:
    1 ml/Ampule
  • Formula:
    C₁₆H₁₀
  • Description:
    Pyrene
  • Cat. No.:
    75835-654
  • Supplier no.:
    31282

 

 

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.