Order Entry
Puerto Rico
ContactUsLinkComponent
Pesticides Mix #2, Method 527 (5 Components), Restek
Pesticides Mix #2, Method 527 (5 Components), Restek
Catalog # 75835-076
Supplier:  Restek
CAS Number:  
Pesticides Mix #2, Method 527 (5 Components), Restek
Catalog # 75835-076
Supplier:  Restek
Supplier Number:  33008
CAS Number:  

Specifications

  • Concentration:
    500 µg/ml each in ethyl acetate
  • Description:
    Pesticides Mix #2, Method 527
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-076
  • Supplier no.:
    33008

Specifications

About this item

The pesticides mix consists of five components chlorpyrifos, dimethoate, malathion, parathion and terbufos sulphone.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.