SOM01.1 VOA Non-Ketone Deuterated Monitoring Compounds, Restek
Catalog # 75834-520
Supplier: Restek
CAS Number:
Specifications
- Concentration:500 µg/ml each in deuterated methanol (MeOD)
- Description:SOM01.1 VOA Non-Ketone Deuterated Monitoring Compounds
- Size:1 ml/Ampule
- Cat. No.:75834-520
- Supplier no.:30624
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.