Order Entry
Puerto Rico
ContactUsLinkComponent
Residual Solvents Class 2 - Mix C (8 Components), Restek
Residual Solvents Class 2 - Mix C (8 Components), Restek
Catalog # 75835-458
Supplier:  Restek
CAS Number:  
Residual Solvents Class 2 - Mix C (8 Components), Restek
Catalog # 75835-458
Supplier:  Restek
Supplier Number:  36273
CAS Number:  

Specifications

  • Concentration:
    In dimethyl sulfoxide
  • Description:
    Residual Solvents Class 2 - Mix C
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-458
  • Supplier no.:
    36273

Specifications

About this item

These mixtures reflect the changes made in USP30/NF25 effective July 1, 2008.

Contains: 2-ethoxyethanol, 800 µg/ml, ethylene glycol, 3100 µg/ml, formamide, 1100 µg/ml, N,N-dimethylacetamide, 5450 µg/ml, N,N-dimethylformamide, 4400 µg/ml, 2-methoxyethanol (methyl cellosolve), 250 µg/ml, N-methylpyrrolidone, 2650 µg/ml, sulfolane, 800 µg/ml.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.