Residual Solvents Class 2 - Mix B (8 Components), Restek
Catalog # 75835-456
Supplier: Restek
CAS Number:
Specifications
- Concentration:In dimethyl sulfoxide
- Description:Residual Solvents Class 2 - Mix B
- Size:1 ml/Ampule
- Cat. No.:75835-456
- Supplier no.:36272
Specifications
About this item
Contains: n-hexane, C6, 1450 µg/ml, chloroform, 300 µg/ml, 1,2-dimethoxyethane, 500 µg/ml, 2-hexanone, 250 µg/ml, nitromethane, 250 µg/ml, pyridine, 1000 µg/ml, tetralin, 500 µg/ml, trichloroethene, 400 µg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.