Standard Mixture of n-alkanes for System Performance Test, Restek
Catalog # 75834-724
Supplier: Restek
CAS Number:
Specifications
- Concentration:50 µg/ml each in cyclohexane
- Description:Standard Mixture of n-alkanes for System Performance Test
- Size:1 ml/Ampule
- Cat. No.:75834-724
- Supplier no.:31633
Specifications
About this item
Reference mixtures for ISO/DIS 9377 (German H-53), a gas chromatography–flame ionization detection (GC-FID) method.
- For GC analysis of total petroleum hydrocarbons (TPH) in water
- Environmentally safer than previous methods
- Calibration standard available as diesel #2/motor oil and diesel #2/mineral oil.
16 components
Certifications: ISO/DIS 9377 Water Quality Testing (German H-53)