Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Atlantic RBCA VPH Mix, Restek
Atlantic RBCA VPH Mix, Restek
Atlantic RBCA VPH Mix, Restek
Catalog # 75834-822
Supplier:
Restek
CAS Number:
Certificates
Expand image
Atlantic RBCA VPH Mix, Restek
Catalog # 75834-822
Supplier:
Restek
Supplier Number:
31871
CAS Number:
Certificates
Specifications
Concentration:
1000 µg/ml each in P and T methanol
Description:
Atlantic RBCA VPH Mix
Size:
1 ml/Ampule
Cat. No.:
75834-822
Supplier no.:
31871
Specifications
About this item
Product Details
Documentation
Product Details
Documentation