Order Entry
Puerto Rico
ContactUsLinkComponent
Carbohydrate LC Performance Check Mix (5 components), Restek
Carbohydrate LC Performance Check Mix (5 components), Restek
Catalog # 75834-770
Supplier:  Restek
CAS Number:  
Carbohydrate LC Performance Check Mix (5 components), Restek
Catalog # 75834-770
Supplier:  Restek
Supplier Number:  31809
CAS Number:  

Specifications

  • Concentration:
    Reconstitute in 1 ml acetonitrile:water (75:25) to 2.1 2.0 4.4 4.5 4.0 mg/ml respectively
  • Description:
    Carbohydrate LC Performance Check Mix
  • Size:
    Dry components in 4 ml screw-cap vial
  • Cat. No.:
    75834-770
  • Supplier no.:
    31809

Specifications

About this item

This carbohydrate LC performance check mix consist of five components which performance qualification (PQ) determines the precision of the LC system.

  • For simple sugar analysis by LC with refractive index/light scattering detection
  • Use with a silica column with amino functionality, as recommended by AOAC International
  • Use as a calibration standard for AACC Method 80-04
  • Offered dry for enhanced stability.

Check mix for LC-RI consists of five simple sugars in varied concentrations: Glucose (2,1 mg), Fructose (2,0 mg), Lactose (4,4 mg), Maltose (4,5 mg), Sucrose (4,0 mg).

Performance qualification (PQ) determines the precision of the LC system. Our performance check mix for LC-RI consists of five simple sugars in varied concentrations. We prepare the reference material in water, dehydrate it, and package it dry for enhanced stability.