Order Entry
Puerto Rico
ContactUsLinkComponent
Fragrance Materials Test Mix (12 Components), Restek
Fragrance Materials Test Mix (12 Components), Restek
Catalog # 75834-768
Supplier:  Restek
CAS Number:  
Fragrance Materials Test Mix (12 Components), Restek
Catalog # 75834-768
Supplier:  Restek
Supplier Number:  31807
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Description:
    Fragrance Materials Test Mix
  • Size:
    0.5 ml in an amber ampule
  • Cat. No.:
    75834-768
  • Supplier no.:
    31807

Specifications

About this item

This fragrance materials test mix idela to performance evaluation for essential oils and fragrance chemicals.

  • Performance evaluation for essential oils and fragrance chemicals
  • System suitability mixture for GC systems and analytical columns
  • Convenient 0.5 mL quantity for easy dilution to recommended working solution

This specially formulated reference standard kit exceeds IFRA purity requirements and contains all 24 EU-defined fragrance allergens detectable by GC-MS, along with seven additional potential allergens. When analyzed on a Restek Rxi-17 column following the IFRA method, all required compounds and isomers resolve clearly with no coelutions. This time-saving mix is also incredibly robust with a shelf life of over one and a half years, so you can stock an already-made standard instead of spending the time to create a new batch for every analysis.

It contain 12 components: Benzoic acid (1,0%), Benzyl salicylate (36,2%), 1,8-Cineole (Eucalyptol) (0,5%), trans Cinnamaldehyde (0,5%), Cinnamyl acetate (0,3%), Cinnamyl alcohol 0,3%, Ethyl butyrate (36,2%), Geraniol (0,6%), Hydroxycitronellal (3,7-Dimethyl-7-hydroxyoctanal) (5,0%), d-Limonene (20,0%), Thymol (0,3%) and Vanilliv (0,1%).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Ordering information: Quantity discounts not available.