Order Entry
Puerto Rico
ContactUsLinkComponent
LC Normal Phase Test Mix #1 (4 Components), Restek
LC Normal Phase Test Mix #1 (4 Components), Restek
Catalog # 75835-328
Supplier:  Restek
CAS Number:  
LC Normal Phase Test Mix #1 (4 Components), Restek
Catalog # 75835-328
Supplier:  Restek
Supplier Number:  35004
CAS Number:  

Specifications

  • Concentration:
    In hexane
  • Description:
    LC Normal Phase Test Mix #1
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-328
  • Supplier no.:
    35004

Specifications

About this item

Routine analysis using this mix can assist in determining the need to perform column and/or system maintenance.

The LC normal phase test mix #1 consists of four components they are benzene (1,00 mg/ml), benzaldehyde, (0,04 mg/ml), benzyl alcohol, (3,00 mg/ml) and 4-methoxybenzyl alcohol (2,00 mg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.