Order Entry
Puerto Rico
ContactUsLinkComponent
FAPAS Series 5 OC Pesticide Mix 2 (19 Components), Restek
FAPAS Series 5 OC Pesticide Mix 2 (19 Components), Restek
Catalog # 75834-960
Supplier:  Restek
CAS Number:  
FAPAS Series 5 OC Pesticide Mix 2 (19 Components), Restek
Catalog # 75834-960
Supplier:  Restek
Supplier Number:  32414
CAS Number:  

Specifications

  • Concentration:
    In acetone
  • Description:
    FAPAS Series 5 OC Pesticide Mix 2
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75834-960
  • Supplier no.:
    32414

Specifications

About this item

Laboratories testing food quality and safety are encouraged to routinely perform proficiency tests. Proficiency testing is an external check of quality. It provides an independent and unbiased assessment of the performance of all aspects of the laboratory, both human and hardware.

  • External check of quality for laboratories performing food testing
  • Ensures accurate proficiency testing
  • Varied concentrations and suitable for GC-ECD analysis

Each participating laboratory is encouraged to use its normal analytical method, thereby simulating the testing of a routine laboratory sample as closely as possible.

Contains 19 components, those are aldrin, α-bhc, β-bhc, γ-bhc, cis-chlordane, trans-chlordane, 4,4'-ddd, 4,4'-dde, 2,4'-ddt, 4,4'-ddt, dieldrin, α-endosulfan, β-endosulfan, endosulfan sulfate, endrin, heptachlor and heptachlor epoxide.

Certifications: Certified reference material (CRM) manufactured and QC-tested in Restek’s ISO-accredited labs—satisfy your ISO requirements.