Minnesota Ag List 1 Pesticides Mix B (6 components), Restek
75834-952
: Restek
:
- Concentration:200 ppm each in acetone
- Description:Minnesota Ag List 1 Pesticides Mix B
- Size:1 ml/Ampule
- Cat. No.:75834-952
- Supplier no.:32407
Contains: Chlorpyrifos, EPTC, fonofos, phorate, terbufos, triallate.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.