ASTM D6042-96 Calibration Mix (7 Components), Restek
75835-724
: Restek
:
- Concentration:50 µg/ml each in isopropanol
- Description:ASTM D6042-96 Calibration Mix
- Size:1 ml/Ampule
- Cat. No.:75835-724
- Supplier no.:31628
This mixture contains the common antioxidants and slips listed in ASTM D6042-96, along with BHT.
The mix consists of BHT, erucamide slip, vitamin E, irgafos 168, irganox 3114, irganox 1010, irganox 1076.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.