Order Entry
Puerto Rico
ContactUsLinkComponent
Certified BTEX in Unleaded Gas Composite Standard (9 Components), Restek
Certified BTEX in Unleaded Gas Composite Standard (9 Components), Restek
Catalog # 75835-548
Supplier:  Restek
CAS Number:  
Certified BTEX in Unleaded Gas Composite Standard (9 Components), Restek
Catalog # 75835-548
Supplier:  Restek
Supplier Number:  30237
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    5500 ppm gasoline in P and T methanol
  • CAS No.:
    8006-61-9
  • Description:
    Certified BTEX in Unleaded Gas Composite Standard
  • Size:
    1 ml/Ampule
  • Description:
    Certified BTEX in Unleaded Gas Composite Standard
  • Cat. No.:
    75835-548
  • Supplier no.:
    30237

Specifications

About this item

The BTEX in unleaded gas composite standard consists of 9 components, they are benzene, ethylbenzene, isopropyl benzene, methyl tert-butyl ether (MTBE), naphthalene, toluene, m-xylene, o-xylene and p-xylene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.