CLP 04.1 Phenols Calibration Mix (14 Components), Restek
Catalog # 75835-690
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Description:CLP 04.1 Phenols Calibration Mix
- Size:1 ml/Ampule
- Cat. No.:75835-690
- Supplier no.:31494
Specifications
About this item
The mix consists of 4-chloro-3-methylphenol, 2-chlorophenol, 2,4-dichlorophenol, 2,4-dimethylphenol, 4,6-dinitro-2-methylphenol, 2,4-dinitrophenol, 2-methylphenol, 4-methylphenol, 2-nitrophenol, 4-nitrophenol, pentachlorophenol, phenol, 2,4,5-trichlorophenol, 2,4,6-trichlorophenol.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.