CLP 04.1 VOA Internal Standard/SMC Spike Mix (6 Components), Restek
Catalog # 75835-566
Supplier: Restek
CAS Number:
Specifications
- Concentration:2500 µg/ml each in P and T methanol
- Description:CLP 04.1 VOA Internal Standard/SMC Spike Mix
- Size:1 ml/Ampule
- Cat. No.:75835-566
- Supplier no.:30457
Specifications
About this item
The VOA internal standard/SMC spike mix consists of 6 components, they are bromochloromethane,1-bromo-4-fluorobenzene (BFB), chlorobenzene-d5, 1,2-dichloroethane-d4, 1,4-difluorobenzene and toluene-d8.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.