$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
8260B Matrix Spike Mix, Restek
8260B Matrix Spike Mix, Restek
75834-394
:
Restek
:
8260B Matrix Spike Mix, Restek
75834-394
:
Restek
:
30479
:
Concentration:
2500 µg/ml each in P and T methanol
Description:
8260B Matrix Spike Mix
Size:
1 ml/Ampule
Cat. No.:
75834-394
Supplier no.:
30479