Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PCB Congener Mix, Method 8082A (19 Components), Restek

PCB Congener Mix, Method 8082A (19 Components), Restek

PCB Congener Mix, Method 8082A, 100 µg/ml each in isooctane, 1 ml/Ampule
Catalog # 75834-962
Supplier:   Restek
Supplier #:  32416
PCB Congener Mix, Method 8082A (19 Components), Restek
Catalog # 75834-962
Supplier:  Restek
CAS Number:  
PCB Congener Mix, Method 8082A (19 Components), Restek

PCB Congener Mix, Method 8082A, 100 µg/ml each in isooctane, 1 ml/Ampule

CAS Number:  
Price
$200.53
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Contains 19 components, those are 2-chlorobiphenyl , 2,3-dichlorobiphenyl , 2,2',5-trichlorobiphenyl , 2,4',5-trichlorobiphenyl, 2,2',3,5'-tetrachlorobiphenyl, 2,2',5,5'-tetrachlorobiphenyl, 2,3',4,4'-tetrachlorobiphenyl, 2,2',3,4,5'-pentachlorobiphenyl, 2,2',4,5,5'-pentachlorobiphenyl, 2,3,3',4',6-pentachlorobiphenyl, 2,2',3,4,4',5'-hexachlorobiphenyl, 2,2',3,4,5,5'-hexachlorobiphenyl, 2,2',3,5,5',6-hexachlorobiphenyl, 2,2',4,4',5,5'-hexachlorobiphenyl, 2,2',3,3',4,4',5-heptachlorobiphenyl, 2,2',3,4,4',5,5'-heptachlorobiphenyl, 2,2',3,4,4',5',6-heptachlorobiphenyl, 2,2',3,4',5,5',6-heptachlorobiphenyl and 2,2',3,3',4,4',5,5',6-nonachlorobiphenyl.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    100 µg/ml each in isooctane
  • Description:
    PCB Congener Mix, Method 8082A
  • Size:
    1 ml/Ampule

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.