Specifications
- Concentration:In methyl tert-butyl ether
- Description:Haloacetic Acid Mix #2
- Size:1 ml/Ampule
- Cat. No.:75835-742
- Supplier no.:31646
Specifications
About this item
The mix consists of, bromochloroacetic acid (400 μg/ml), bromodichloroacetic acid (400 μg/ml), chlorodibromoacetic acid (1000 μg/ml), dibromoacetic acid (200 μg/ml), dichloroacetic acid (600 μg/ml), monobromoacetic acid (400 μg/ml), monochloroacetic acid (600 μg/ml), tribromoacetic acid (2000 μg/ml), trichloroacetic acid (200 μg/ml)
- Prepared from purified neat reference materials for the highest quality
- Lot-to-lot consistency ensures analytical reproducibility