ContactUsLinkComponent
EPA 539 Calibration Stock Standard (7 Components), Restek
EPA 539 Calibration Stock Standard (7 Components), Restek
  75834-918
 :  Restek
 :  
EPA 539 Calibration Stock Standard (7 Components), Restek
  75834-918
 :  Restek
 :  31998
 :  
  • Matrix:
    Acetonitrile
  • Description:
    EPA 539 Calibration Stock Standard
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75834-918
  • Supplier no.:
    31998
Contains the seven hormones listed in EPA Method 539 at the appropriate concentrations. The seven components are 4-androstene-3, 17-dione, equilin, 17-beta-Estradiol, estriol, estrone, 17-alpha-ethynylestradiol and testosterone.
 : Contains the seven hormones listed in EPA Method 539 at the appropriate concentrations. The seven components are 4-androstene-3, 17-dione, equilin, 17-beta-Estradiol, estriol, estrone, 17-alpha-ethynylestradiol and testosterone.
 : UCMR3 method 539 calibration standard with concentrations of 10 to 400 µg/ml to more conveniently match UCMR3 requirements (cat.# 32461) also available.