Canadian Drinking Water Phenoxyacid Herbicides Mix (11 Components), Restek
Catalog # 75834-820
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each
- Matrix:Acetone
- Description:Canadian Drinking Water Phenoxyacid Herbicides Mix
- Size:1 ml/Ampule
- Cat. No.:75834-820
- Supplier no.:31868
Specifications
About this item
Consists of 11 components, those are bromoxynil, 2,4-d, dicamba, 2,4-dichlorophenol, diclofop methyl, dinoseb, pentachlorophenol, picloram, 2,4,5-t, 2,3,4,6-tetrachlorophenol and 2,4,6-trichlorophenol.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.