Order Entry
Puerto Rico
ContactUsLinkComponent
CLP 04.1 BNA Surrogate Mix (8 Components), Restek
CLP 04.1 BNA Surrogate Mix (8 Components), Restek
Catalog # 75835-688
Supplier:  Restek
CAS Number:  
CLP 04.1 BNA Surrogate Mix (8 Components), Restek
Catalog # 75835-688
Supplier:  Restek
Supplier Number:  31493
CAS Number:  

Specifications

  • Matrix:
    Methylene chloride
  • Description:
    CLP 04.1 BNA Surrogate Mix
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-688
  • Supplier no.:
    31493

Specifications

About this item

The mix consists of components such as 2-chlorophenol-d4 (1500 μg/ml), 1,2-dichlorobenzene-d4 (1000 μg/ml), 2-fluorobiphenyl (1000 μg/ml), 2-fluorophenol (1500 μg/ml), nitrobenzene-d5 (1000 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.