Order Entry
Puerto Rico
ContactUsLinkComponent
California Oxygenates Mix (5 components), Restek
California Oxygenates Mix (5 components), Restek
Catalog # 75835-572
Supplier:  Restek
CAS Number:  
California Oxygenates Mix (5 components), Restek
Catalog # 75835-572
Supplier:  Restek
Supplier Number:  30465
CAS Number:  

Specifications

  • Matrix:
    Methanol (P&T)
  • Description:
    California Oxygenates Mix
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-572
  • Supplier no.:
    30465

Specifications

About this item

The california oxygenates mix consists of five components they are tert-amyl methyl ether (TAME) 2000 μg/ml, tert-butanol (TBA) 10000 μg/ml, diisopropyl ether (DIPE) 2000 μg/ml, ethyl-tert-butyl ether (ETBE) 2000 μg/ml and methyl tert-butyl ether (MTBE) 2000 μg/ml.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.