Sample Tubes, Polypropylene, PerkinElmer
PP, 50 ml, 28 × 115 mm
Catalog # 97012-780
Supplier: PerkinElmer
Supplier #: B0193234
Jump to:
Product Details & Documents
Translucent polypropylene conical freestanding tubes are made to provide the excellent chemical resistance.
- Graduation
Specifications
- Volume:50 ml
- Material:PP
- Size:28 × 115 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



