Order Entry
Puerto Rico
ContactUsLinkComponent
Diesel #2/Motor Oil (2 Components), Restek
Diesel #2/Motor Oil (2 Components), Restek
  75834-736
 :  Restek
 :  
Diesel #2/Motor Oil (2 Components), Restek
  75834-736
 :  Restek
 :  31682
 :  

 

  • Concentration:
    5000 µg/ml each in hexane
  • Description:
    Diesel #2/Motor Oil
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75834-736
  • Supplier no.:
    31682

 

 

Diesel fuel #2 composite and Motor oil.

It has two components.

Shelf life 6 to 85 months.