Order Entry
Puerto Rico
ContactUsLinkComponent
ASTM D2887-12 Calibration Standard (20 Components), Restek
ASTM D2887-12 Calibration Standard (20 Components), Restek
Catalog # 75835-778
Supplier:  Restek
CAS Number:  
ASTM D2887-12 Calibration Standard (20 Components), Restek
Catalog # 75835-778
Supplier:  Restek
Supplier Number:  31674
CAS Number:  

Specifications

  • Concentration:
    1% w/w in carbon disulfide
  • Description:
    ASTM D2887-12 Calibration Standard
  • Size:
    1 g solution/ampule
  • Cat. No.:
    75835-778
  • Supplier no.:
    31674

Specifications

About this item

Designed to serve as a column resolution text mixture, as well as a calibration mixture for retention time (RT) versus boiling point (bp) calibration.

The mix consists of, (C5) n-pentane, (C6) n-hexane, (C7) n-heptane, (C8) n-octane, (C9) n-nonane, (C10) n-decane, (C11) n-undecane, (C12) n-dodecane, (C14) n-tetradecane, (C15) n-pentadecane, (C16) n-hexadecane, (C17) n-heptadecane, (C18) n-octadecane, (C20) n-eicosane, (C24) n-tetracosane, (C28) n-octacosane, (C32) n-dotriacontane, (C36) n-hexatriacontane, (C40) n-tetracontane, (C44) n-tetratetracontane.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.