Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Michigan GRO Mix (14 Components), Restek

Michigan GRO Mix (14 Components), Restek

Michigan GRO Mix, 2000 µg/ml each in P and T methanol, 1 ml, Ampoule
Catalog # 75835-578
Supplier:   Restek
Supplier #:  30468
Michigan GRO Mix (14 Components), Restek
Catalog # 75835-578
Supplier:  Restek
CAS Number:  
Michigan GRO Mix (14 Components), Restek

Michigan GRO Mix, 2000 µg/ml each in P and T methanol, 1 ml, Ampoule

CAS Number:  
Price
$57.85
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

It consists of benzene, 1,2-dibromoethane, 1,2-dichloroethane, ethylbenzene, isopropylbenzene (cumene), 2-methylnaphthalene, methyl tert-butyl-ether (MTBE), naphthalene, toluene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene and p-xylene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.

Specifications

Specifications

  • Concentration:
    2000 µg/ml each in P and T methanol
  • Pack type:
    Ampoule
  • Description:
    Michigan GRO Mix
  • Size:
    1 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.