MA Fractionation Check Mix (31 Components), Restek
75834-718
: Restek
:
- Concentration:25 µg/ml each in hexane
- Pack type:Ampoule
- Description:MA Fractionation Check Mix
- Size:1 ml
- Cat. No.:75834-718
- Supplier no.:31481
This MA fractionation check mix ideal for underground storage tank monitoring.
It contains 31 components:
PAHs: Acenaphthene, Acenaphthylene, Anthracene, Benz(a)anthracene, Benzo(a)pyrene, Benzo(b)fluoranthene, Benzo(k)fluoranthene, Benzo(ghi)perylene, Chrysene, Dibenz(a,h)anthracene, Fluoranthene, Fluorene, Indeno(1,2,3-cd)pyrene, 2-Methylnaphthalene, Naphthalene, Phenanthrene and Pyrene.
Hydrocarbons: (C9) n-Nonane, (C10) n-Decane, (C12) n-Dodecane, (C14) n-Tetradecane, (C16) n-Hexadecane, (C18) n-Octadecane, (C19) n-Nonadecane, (C20) n-Eicosane, (C22) n-Docosane, (C24) n-Tetracosane, (C26) n-Hexacosane, (C28) n-Octacosane, (C30) n-Triacontane and (C36) n-Hexatriacontane
Shelf life 6 months.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.