Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Caspase 7
Human Recombinant Caspase 7
Catalog # 82022-064
Supplier:  G-Biosciences
CAS Number:  
Human Recombinant Caspase 7
Catalog # 82022-064
Supplier:  G-Biosciences
Supplier Number:  RCH-007
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Species:
    Human
  • Size:
    25 units
  • Enzyme Name:
    Caspase 7
  • Cat. No.:
    82022-064
  • Supplier no.:
    RCH-007

Specifications

About this item

G-Biosciences' Recombinant Human Caspases 1-3 and 6-10 are supplied lyophilized in quantities of 25 units

G-Biosciences' Recombinant Human Caspases 1-3 and 6-10 are members of the ICE (interleukin-1 β converting enzyme) family of cysteine proteases. They exist in cells as inactive pro-enzymes. During apoptosis, pro-caspases 1-3 and 6-10 are processed at aspartate residues by self-proteolysis and/or cleavage by another caspase. The matured (processed) form of these caspases consists of large and small subunits, which associate to form a heterotetramer active enzyme.

G-Biosciences' active recombinant caspase-2 cleaves caspase-2 chromogenic substrate Ac-VDVAD-pNA, and the signal can be detected spectrophotometrically.

G-Biosciences' active caspase-6 and caspase-7 have been shown in the involvement of the proteolysis of poly (ADP-ribose) polymerase (PARP), an enzyme that is involved in DNA repair and genomic maintenance. The active recombinant caspase-6 cleaves the substrates consisting of consensus sequence VEID (e.g. VEID-AFC and VEID-pNA).

One unit of the specific recombinant caspase is the enzyme activity that cleaves one nanomole of a specific caspase substrate per hour at 37°C in a reaction solution containing 50mM HEPES (at a pH of 7.2), 50mM NaCl, 0.1% CHAPS, 10mM EDTA, 5% Glycerol, and 10mM DTT.

Ordering information: Accessories available