Order Entry
Puerto Rico
ContactUsLinkComponent
Mlu I
Mlu I
Catalog # PAR6381
CAS Number:  
undefined
Mlu I
Catalog # PAR6381
Supplier Number:  R6381
CAS Number:  

Specifications

  • Description:
    Mlu I
  • Size:
    1000 U
  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Mlu I
  • Buffer supplied with enzyme:
    Buffer D
  • Buffer activity:
    % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 25…50% – C: 50…75% – D: 1; MULTI-CORE™ Buffer 10…25%
  • Enzyme assay temperature:
    37°C
  • Cat. No.:
    PAR6381
  • Supplier no.:
    R6381

Specifications

About this item

A/CGCG T T GCGC\A

  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications