XbaI
Supplier: Promega Corporation
About this item
T/CTAG A A GATC\T
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Genome qualified to ensure optimal performance in genomic analysis applications
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
T/CTAG A A GATC\T
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Genome qualified to ensure optimal performance in genomic analysis applications
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- PAR6181
- PAR6185
- Enzyme Name:
- XbaI
- XbaI
- Buffer supplied with enzyme:
- Buffer D
- Buffer D
- Buffer activity:
- % Activity (37°C) in 4 CORE® Buffers A: 50…75% – B: 75…100% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 1
- % Activity (37°C) in 4 CORE® Buffers A: 50…75% – B: 75…100% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 1
- Enzyme assay temperature:
- 37°C
- 37°C
- Storage Conditions:
- –30...–10 °C
- –30...–10 °C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- PAR6181XbaISupplier Number: R6181Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.PAR6181Enzyme NameXbaIBuffer supplied with enzymeBuffer DBuffer activity% Activity (37°C) in 4 CORE® Buffers A: 50…75% – B: 75…100% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 1Enzyme assay temperature37°CStorage Conditions–30...–10 °C - PAR6185XbaISupplier Number: R6185Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.PAR6185Enzyme NameXbaIBuffer supplied with enzymeBuffer DBuffer activity% Activity (37°C) in 4 CORE® Buffers A: 50…75% – B: 75…100% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 1Enzyme assay temperature37°CStorage Conditions–30...–10 °C



