SmaI, Promega
Supplier: Promega Corporation
Certificates
About this item
CCC/GGG GGG\CCC
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Genome qualified to ensure optimal performance in genomic analysis applications
- Available at high concentration, containing 5,000 units of SmaI at a concentration of 40-80u/microl
Specifications
- Storage Conditions:–30...–10 °C
- Enzyme Name:SmaI
- Buffer supplied with enzyme:Buffer J
- Buffer activity:% Activity (25°C) in 4 CORE® Buffers A: < 10% – B: < 10% – C: 0,1 – D: < 10%; MULTI-CORE™ Buffer 1
- Enzyme assay temperature:25°C