Order Entry
Puerto Rico
ContactUsLinkComponent
EcoR V (Blue/White Cloning Qualified), Restriction Enzymes, Promega
EcoR V (Blue/White Cloning Qualified), Restriction Enzymes, Promega
Certificates
undefined
EcoR V (Blue/White Cloning Qualified), Restriction Enzymes, Promega
Certificates

Specifications

  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    EcoR V (Blue/White Cloning Qualified)
  • Buffer supplied with enzyme:
    Buffer D
  • Buffer activity:
    % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 25…50% – C: 50…75% – D: 1; MULTI-CORE™ Buffer 1
  • Enzyme assay temperature:
    37°C

Specifications

Related Information