Order Entry
Puerto Rico
ContactUsLinkComponent
 
Hind III
 
undefined
Hind III
 

 

  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Hind III
  • Buffer supplied with enzyme:
    Buffer E
  • Buffer activity:
    % Activity (37°C) in 4 CORE® Buffers A: 25…50% – B: 1 – C: 75…100% – D: 10…25%; MULTI-CORE™ Buffer 50…75%
  • Enzyme assay temperature:
    37°C