ContactUsLinkComponent
EZLabel™ Protein Cy5 Labeling Kit, BioVision
EZLabel™ Protein Cy5 Labeling Kit, BioVision
  75837-056
 :  Avantor
 :  
undefined
EZLabel™ Protein Cy5 Labeling Kit, BioVision
  75837-056
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Host:
  • Description:
    EZLabel™ Protein Cy5 Labeling Kit
  • Environmentally Preferable:
  • Cat. No.:
    75837-056
  • Supplier no.:
    K839-5
Easy to use Kit to label upto 5 mg of Protein wz Cy5 for multiple applications such as ELISA, wblot.
  • All necessary reagents included.
  • Fast and efficient labeling of proteins with Cy5
  • Easy to use Kit to label upto 5 mg of Protein with Cy5 for multiple applications such as ELISA, western blot, Immunohistochemistry, Immunofluorescence and FACS analysis
BioVision’s EZLabel™ Protein-Cy5 Labeling Kit provides an easy way to label proteins with Cy5 in a user-friendly spin column format. Cy5 is an ideal fluorescent dye for labeling of proteins with its intense signal in the red region of the spectrum. Each pair of spin columns provided in the kit can be used to purify up to 1 mg of the labeled target protein. The kit provides all of the reagents necessary to perform five labeling reactions using up to 1 mg of protein per reaction. The Cy5-labeled protein has excitation and emission wavelengths at 650 nm and 670 nm respectively, and can be directly used for downstream applications including ELISA, western blot, Immunohistochemistry, Immunofluorescence, and FACS, etc.