PEEK Ferrules, MicroSolv
Double Cone Ferrule, PEEK, ¹⁄₁₆"
Catalog # 97055-862
Supplier: MicroSolv Technology Corp
Supplier #: 47320-35
Jump to:
Product Details & Documents
Double cone ferrules for use with 2 piece male nuts.
Specifications
- Description:Double Cone Ferrule
- Material:PEEK
- Ferrule size:¹⁄₁₆"