Order Entry
Puerto Rico
ContactUsLinkComponent
 
Digoxigenin-dUTP, Alkali-Stable, Enzo Life Sciences
  10842-814
 :  
undefined
Digoxigenin-dUTP, Alkali-Stable, Enzo Life Sciences
  10842-814
 :  NUC113-0025
 :  

 

  • Description:
    Digoxigenin-dUTP, alkali-stable
  • Size:
    25 nmol
  • Storage Conditions:
    -20°C
  • Shipping Conditions:
    Shipped on Dry Ice
  • Purity:
    ≥93% (HPLC)
  • Concentration:
    1 mM
  • Formulation:
    Liquid. Solution in water.
  • Cat. No.:
    10842-814
  • Supplier no.:
    NUC113-0025

 

 

Digoxigenin-labeled nucleotide suitable for non-radioactive labeling of DNA probes.

  • Replaces dTTP in reactions in which it serves as a substrate for E. coli DNA polymerase
  • Produce digoxigenin-labeled DNA probes in a variety of labeling reactions

Digoxigenin-dUTP replaces dTTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase. Digoxigenin-dUTP can be used to produce digoxigenin-labeled DNA probes in a variety of labeling reactions including nick translation, random prime labeling, cDNA labeling and 3’-end labeling. The resulting digoxigenin-labeled probe can be used in a variety of hybridization applications incuding Southern blots, Northern blots, or dot blots. The probes can also be used for in situ hybridization procedures on fixed cells and tissues. Digoxigenin-labeled probes have been shown to hybridize to homologous nucleic acid at the same rate and to the same extent as non-labeled probes. The hybridized digoxigenin-labeled DNA probes can be detected by their interaction with antibodies coupled to fluorescent dyes or color-producing enzymes.