Methotrexate ELISA Kit, Enzo Life Sciences
Catalog # 75786-344
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Description:Methotrexate ELISA Kit
- Size:1 x 96 wells
- Sensitivity:0.087 ng/ml (0.13-1000 ng/ml)
- Cat. No.:75786-344
- Supplier no.:KIT142-0001
Specifications
About this item
Ultra sensitive Methotrexate ELISA kit enabling detection of Methotrexate in serum, plasma, and urine in just 1.5 hours.
- Sensitive measurement of methotrexate, detecting as little as 0.087 ng/ml
- Broad dynamic range suitable for a large variety of samples
- High throughput format with results in 1.5 hours for up to 38 samples in duplicate
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of methotrexate in serum, plasma and urine in human, mouse, and rat samples.
The Methotrexate ELISA kit is a colorimetric competitive immunoassay kit with results in 1.5 hours.
Ordering information: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.