ContactUsLinkComponent
TGF-alpha ELISA Kit, Enzo Life Sciences
  75786-644
 :  
TGF-alpha ELISA Kit, Enzo Life Sciences
  75786-644
 :  KIT132-0001
 :  
  • Description:
    TGF-α ELISA Kit
  • Size:
    1 x 96 wells
  • Sensitivity:
    9.375 pg/ml
  • Cat. No.:
    75786-644
  • Supplier no.:
    KIT132-0001
Highly sensitive ELISA kit enabling detection of TGF-α.
  • Sensitive measurement of TGF-α, detecting as little as 9.375 pg/ml
  • Negligible reactivity from similar growth factors
  • High throughput format with results in 3.5 hours
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of TGF-α in serum, plasma, cell culture supernatant, tissue homogenates in human samples.
The TGF-α ELISA kit is a colorimetric immunometric immunoassay kit.
 : Contains Microtiter Plate, Standard, Detection Antibody, Detection Antibody Diluent, Sample Diluent, Wash Buffer Concentrate, Conjugate, Conjugate Diluent, Substrate, and Stop Solution.