TSH ELISA Kit, Enzo Life Sciences
75786-642
:
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:HRP-Streptavidin
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Thyroid Stimulating Hormone
- Description:TSH ELISA Kit
- Size:1 x 96 wells
- Storage Temperature:2…8 °C
- Application:Thyroid Function
- Sensitivity:0.5 µIU/ml (0.5-40 µIU/ml)
- Cat. No.:75786-642
- Supplier no.:KIT131-0001
Highly sensitive TSH ELISA kit enabling detection of TSH in serum.
- Sensitive measurement of TSH, detecting as little as 0.5 µIU/ml
- Negligible reactivity from similar glycoprotein hormones
- High throughput format with results in 75 minutes
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of TSH in serum in human samples.
The TSH ELISA kit is a colorimetric immunometric immunoassay kit.
: Contains Microtiter Plate, Standard, Wash Buffer Concentrate, Conjugate, Substrate, and Stop Solution.